Gene Transcripts
- Gene Symbol
- Lrrc8c
- Entrez Gene ID
- 100604
- Gene Name
- leucine rich repeat containing 8 family, member C
- Gene Aliases
- AD158, AI326115, E430036I04Rik, fad158
- Chromosome Location
- Chr. 5: 105519386 - 105613018 on Build GRCm38
- UniGene ID
- Mm.319847
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK081556.1 |
- |
1 |
2374 |
|
AK168156.1 |
- |
1 |
1062 |
|
BC050074.1 |
- |
1 |
208 |
|
View Full Product Details