Gene Transcripts
- Gene Symbol
- MGAT2
- Entrez Gene ID
- 4247
- Gene Name
- mannosyl (alpha-1,6-)-glycoprotein beta-1,2-N-acetylglucosaminyltransferase
- Gene Aliases
- CDG2A, CDGS2, GLCNACTII, GNT-II, GNT2
- Chromosome Location
- Chr. 14: 49620771 - 49623481 on Build GRCh38
- UniGene ID
- Hs.93338
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BQ023756.1 |
- |
1 |
320 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000555043.1 |
- |
1 |
303 |
|
Chr. 14 - 49620815 - 49623480 [-] |
View Full Product Details