Gene Transcripts
- Gene Symbol
- KCNJ14
- Entrez Gene ID
- 3770
- Gene Name
- potassium voltage-gated channel subfamily J member 14
- Gene Aliases
- IRK4, KIR2.4
- Chromosome Location
- Chr. 19: 48455707 - 48466110 on Build GRCh38
- UniGene ID
- Hs.144011
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BU739644.1 |
- |
1 |
393 |
|
- Gene Symbol
- LOC105372430
- Entrez Gene ID
- 105372430
- Gene Name
- uncharacterized LOC105372430
- Gene Aliases
- Chromosome Location
- Chr. 19: 48465471 - 48469507 on Build GRCh38
- UniGene ID
- Hs.736972
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC042033.1 |
- |
3 |
512 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000593476.1 |
- |
3 |
511 |
|
Chr. 19 - 48465611 - 48469693 [-] |
GENCODE |
ENST00000600529.1 |
- |
2 |
577 |
|
Chr. 19 - 48465593 - 48466937 [-] |
View Full Product Details