Gene Transcripts

Gene Symbol
CNTN3
Entrez Gene ID
5067
Gene Name
contactin 3
Gene Aliases
BIG-1, PANG, PCS
Chromosome Location
Chr. 3: 74262568 - 74614448 on Build GRCh38
UniGene ID
Hs.12723
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020872.2 NP_065923.1 13 1714
XM_005264757.3 XP_005264814.1 14 1824
XM_011533768.2 XP_011532070.1 13 5346
XM_017006507.1 XP_016861996.1 14 2394
XM_017006508.1 XP_016861997.1 13 1619
XM_017006509.1 XP_016861998.1 4 393
GenBank mRNA AB040929.1 - 11 1363
BC150608.1 - 13 1714 9056978

View Full Product Details