Gene Transcripts

Gene Symbol
Trp53bp2
Entrez Gene ID
209456
Gene Name
transformation related protein 53 binding protein 2
Gene Aliases
53BP2, AI746547, ASPP2, PPP1R13A, Tp53bp2, X98550
Chromosome Location
Chr. 1: 182409167 - 182462436 on Build GRCm38
UniGene ID
Mm.287450
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173378.2 NP_775554.2 13 2809
GenBank mRNA AK134556.1 - 6 1779
BC030894.1 - 5 1438
BC042874.1 - 12 2411

View Full Product Details