Gene Transcripts
- Gene Symbol
- Wdr26
- Entrez Gene ID
- 226757
- Gene Name
- WD repeat domain 26
- Gene Aliases
- 1600024A01Rik, AA693241, AI447817, AU044014, C77982, Gid7
- Chromosome Location
- Chr. 1: 181173226 - 181211978 on Build GRCm38
- UniGene ID
- Mm.289082
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_145514.5 |
NP_663489.4
|
8, 9 |
1783 |
|
GenBank mRNA |
AK075743.1 |
- |
8, 9 |
1175 |
|
AK082326.1 |
- |
9, 10 |
1078 |
|
AK194190.1 |
- |
1, 2 |
116 |
|
AK194610.1 |
- |
1, 2 |
116 |
|
AK194679.1 |
- |
1, 2 |
116 |
|
AK195392.1 |
- |
1, 2 |
116 |
|
AK201026.1 |
- |
1, 2 |
116 |
|
BC020044.1 |
- |
4, 5 |
528 |
|
BC058601.1 |
- |
6, 7 |
779 |
|
BC094233.1 |
- |
5, 6 |
667 |
|
BC139336.1 |
- |
8, 9 |
945 |
8862358 |
BC139337.1 |
- |
8, 9 |
945 |
8862359 |
View Full Product Details