Gene Transcripts

Gene Symbol
Slc13a4
Entrez Gene ID
243755
Gene Name
solute carrier family 13 (sodium/sulfate symporters), member 4
Gene Aliases
9630060C05Rik, SUT-1, SUT1
Chromosome Location
Chr. 6: 35267953 - 35308126 on Build GRCm38
UniGene ID
Mm.23666
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172892.3 NP_766480.1 10 1774
GenBank mRNA AK036354.1 - 10 1776
AK038937.1 - 10 1779
BC089161.1 - 10 1763 30440173

View Full Product Details