Gene Transcripts

Gene Symbol
Armcx6
Entrez Gene ID
278097
Gene Name
armadillo repeat containing, X-linked 6
Gene Aliases
AW060994
Chromosome Location
Chr. X: 134748447 - 134756342 on Build GRCm38
UniGene ID
Mm.113272
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007578.2 NP_001007579.1 2 104
GenBank mRNA AK148507.1 - 2 103
BC027264.1 - 2 97 3492516
BY777182.1 - 2 84

View Full Product Details