Gene Transcripts

Gene Symbol
Upf2
Entrez Gene ID
326622
Gene Name
UPF2 regulator of nonsense transcripts homolog (yeast)
Gene Aliases
Chromosome Location
Chr. 2: 5951447 - 6056703 on Build GRCm38
UniGene ID
Mm.31102
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081132.1 NP_001074601.1 13 2603
GenBank mRNA BC039278.1 - 2 266
BC138358.1 - 13 2566 8861378
BC138359.1 - 13 2566 8861379

View Full Product Details