Gene Transcripts

Gene Symbol
Khdc1a
Entrez Gene ID
368204
Gene Name
KH domain containing 1A
Gene Aliases
AU014713, C88118, Ndg1
Chromosome Location
Chr. 1: 21329476 - 21352200 on Build GRCm38
UniGene ID
Mm.26006
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183322.2 NP_899145.2 3 1621
GenBank mRNA AK162365.1 - 1 717
AY238603.1 - 3 1384

View Full Product Details