Gene Transcripts
- Gene Symbol
- Khdc1a
- Entrez Gene ID
- 368204
- Gene Name
- KH domain containing 1A
- Gene Aliases
- AU014713, C88118, Ndg1
- Chromosome Location
- Chr. 1: 21329476 - 21352200 on Build GRCm38
- UniGene ID
- Mm.26006
- Species
- Mus musculus
- Gene Symbol
- Khdc1c
- Entrez Gene ID
- 433278
- Gene Name
- KH domain containing 1C
- Gene Aliases
- Khdc1b
- Chromosome Location
- Chr. 1: 21368331 - 21369743 on Build GRCm38
- UniGene ID
- Mm.26006
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC141037.1 |
- |
|
1152 |
|
View Full Product Details