Gene Transcripts

Gene Symbol
Prickle4
Entrez Gene ID
381104
Gene Name
prickle homolog 4 (Drosophila)
Gene Aliases
Gm939
Chromosome Location
Chr. 17: 47688474 - 47694958 on Build GRCm38
UniGene ID
Mm.392206
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290337.1 NP_001277266.1 4 601
GenBank mRNA BX525727.1 - 2 170

View Full Product Details