Gene Transcripts

Gene Symbol
Odf3l1
Entrez Gene ID
382075
Gene Name
outer dense fiber of sperm tails 3-like 1
Gene Aliases
BC049697, Gm1116
Chromosome Location
Chr. 9: 56848657 - 56864674 on Build GRCm38
UniGene ID
Mm.335177
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198673.2 NP_941075.1 5 576
GenBank mRNA AK142219.1 - 6 711
BC049697.1 - 5 576 6706514

View Full Product Details