Gene Transcripts

Gene Symbol
Mrfap1
Entrez Gene ID
67568
Gene Name
Morf4 family associated protein 1
Gene Aliases
9130413I22Rik, AL022827, Pam14
Chromosome Location
Chr. 5: 36794867 - 36796754 on Build GRCm38
UniGene ID
Mm.332336
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026242.3 NP_080518.1 2 1431
GenBank mRNA AB041651.1 - 2 1411
AK005313.1 - 2 1423
AK017832.1 - 2 1413
AK018675.1 - 2 1381
AK170789.1 - 2 1381
AK170909.1 - 2 1409
AK170984.1 - 2 1413
AK211332.1 - 1 235
BC010209.1 - 2 1400 3482091
BC085488.1 - 2 1276

View Full Product Details