Gene Transcripts

Gene Symbol
Trav7n-5
Entrez Gene ID
667635
Gene Name
T cell receptor alpha variable 7N-5
Gene Aliases
EG667635, Gm8740
Chromosome Location
Chr. 14: 53146922 - 53147448 on Build GRCm38
UniGene ID
Mm.489210
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC064122.1 - 1471

View Full Product Details