Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012425.3 | NP_036557.1 | 4 | 394 | |
NM_152724.2 | NP_689937.2 | 3 | 282 | ||
XM_005252552.3 | XP_005252609.1 | 4 | 477 | ||
GenBank mRNA | AK225681.1 | - | 4 | 336 | |
AK292911.1 | - | 3 | 207 | ||
AK312520.1 | - | 4 | 337 | ||
BC005993.1 | - | 4 | 364 | 4050837 | |
BC008691.1 | - | 3 | 553 | 3872071 | |
BC015644.2 | - | 4 | 354 | 4647836 | |
BC107898.1 | - | 3 | 261 | ||
BG400752.1 | - | 3 | 283 | ||
BX647733.1 | - | 3 | 258 | ||
CR536521.1 | - | 3 | 251 | ||
CR541840.1 | - | 3 | 251 |