Gene Transcripts

Gene Symbol
Myh7b
Entrez Gene ID
668940
Gene Name
myosin, heavy chain 7B, cardiac muscle, beta
Gene Aliases
Myh14
Chromosome Location
Chr. 2: 155611243 - 155634307 on Build GRCm38
UniGene ID
Mm.469980
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085378.2 NP_001078847.1 15 1458

View Full Product Details