Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001130440.1 | NP_001123912.1 | 1 | 171 | |
NM_003133.5 | NP_003124.1 | 1 | 171 | ||
GenBank mRNA | AF070649.1 | - | 1 | 150 | |
AK289595.1 | - | 1 | 136 | ||
AL050363.1 | - | 1 | 119 | ||
BC008443.1 | - | 1 | 155 | 4102222 | |
BC015094.1 | - | 1 | 142 | 3930047 | |
BC021995.1 | - | 1 | 151 | 4101294 | |
BC022415.1 | - | 1 | 137 | 4243562 | |
BC064351.1 | - | 1 | 120 | 4452218 | |
BC066957.1 | - | 1 | 158 | 5263635 | |
BC067845.1 | - | 1 | 106 | 5287027 | |
BC106005.1 | - | 1 | 147 | 6026221 | |
DA983529.1 | - | 1 | 171 | ||
DB194289.1 | - | 1 | 176 | ||
EF488978.1 | - | 1 | 155 | ||
U20998.1 | - | 1 | 149 |