Gene Transcripts

Gene Symbol
THBS2
Entrez Gene ID
7058
Gene Name
thrombospondin 2
Gene Aliases
TSP2
Chromosome Location
Chr. 6: 169215780 - 169254114 on Build GRCh38
UniGene ID
Hs.371147
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003247.3 NP_003238.2 22 3740
GenBank mRNA AK292429.1 - 22 3697
BC146676.1 - 21 3547 40148868
BC150175.1 - 21 3551 40147958
BX641023.1 - 8 1195
L12350.1 - 22 3659
M81339.1 - 11 1744

View Full Product Details