Gene Transcripts

Gene Symbol
Dnajc15
Entrez Gene ID
290370
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 15
Gene Aliases
Dnajd1
Chromosome Location
Chr. 15: 59990131 - 60056710 on Build Rnor_6.0
UniGene ID
Rn.8092
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106050.1 NP_001099520.1 5, 6 542
GenBank mRNA FQ229062.1 - 5, 6 416

View Full Product Details