Gene Transcripts

Gene Symbol
Erf
Entrez Gene ID
292721
Gene Name
Ets2 repressor factor
Gene Aliases
Chromosome Location
Chr. 1: 82112449 - 82120902 on Build Rnor_6.0
UniGene ID
Rn.9093
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170335.1 NP_001163806.1 4 1008
XM_008758916.1 XP_008757138.1 4 1924
XM_008758917.1 XP_008757139.1 4 1774

View Full Product Details