Gene Transcripts

Gene Symbol
Olr19
Entrez Gene ID
293081
Gene Name
olfactory receptor 19
Gene Aliases
Chromosome Location
Chr. 1: 149913647 - 149914606 on Build Rnor_6.0
UniGene ID
Rn.141949
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000117.1 NP_001000117.1 1 75

Gene Symbol
LOC100909935
Entrez Gene ID
100909935
Gene Name
olfactory receptor 6F1-like
Gene Aliases
Chromosome Location
Chr. 1: 149777726 - 149778712 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003748915.2 XP_003748963.1 1 86

Gene Symbol
LOC100910993
Entrez Gene ID
100910993
Gene Name
olfactory receptor 6F1-like
Gene Aliases
Chromosome Location
Chr. 1|NW_007906107: 2041 - 3114 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003752171.3 XP_003752219.1 1 144

Gene Symbol
Olr17
Entrez Gene ID
405261
Gene Name
olfactory receptor 17
Gene Aliases
Chromosome Location
Chr. 1: 149682401 - 149683357 on Build Rnor_6.0
UniGene ID
Rn.141950
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000937.1 NP_001000937.1 72

View Full Product Details