Gene Transcripts
- Gene Symbol
- ZNF33A
- Entrez Gene ID
- 7581
- Gene Name
- zinc finger protein 33A
- Gene Aliases
- KOX2, KOX31, KOX5, NF11A, ZNF11, ZNF11A, ZNF33, ZZAPK
- Chromosome Location
- Chr. 10: 38010483 - 38067834 on Build GRCh38
- UniGene ID
- Hs.435774
- Species
- Homo sapiens
- Gene Symbol
- ZNF33B
- Entrez Gene ID
- 7582
- Gene Name
- zinc finger protein 33B
- Gene Aliases
- KOX2, KOX31, ZNF11B
- Chromosome Location
- Chr. 10: 42574169 - 42638837 on Build GRCh38
- UniGene ID
- Hs.435774
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK025638.1 |
- |
1 |
136 |
|
View Full Product Details