Gene Transcripts

Gene Symbol
Efhc2
Entrez Gene ID
302507
Gene Name
EF-hand domain (C-terminal) containing 2
Gene Aliases
RGD1563388
Chromosome Location
Chr. X: 5825781 - 6026144 on Build Rnor_6.0
UniGene ID
Rn.39302
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106952.1 NP_001100422.1 11 1824

View Full Product Details