Gene Transcripts
- Gene Symbol
- Telo2
- Entrez Gene ID
- 302986
- Gene Name
- telomere maintenance 2
- Gene Aliases
- RGD1305553
- Chromosome Location
- Chr. 10: 14461581 - 14476808 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_220236.7 |
XP_220236.3
|
8 |
1334 |
|
View Full Product Details