Gene Transcripts

Gene Symbol
Anln
Entrez Gene ID
363031
Gene Name
anillin, actin binding protein
Gene Aliases
RGD1561367
Chromosome Location
Chr. 8: 23336455 - 23397115 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008766019.1 XP_008764241.1 17, 18 2903
XM_008766020.1 XP_008764242.1 17, 18 2903

Gene Symbol
Anlnl1
Entrez Gene ID
307056
Gene Name
anillin, actin binding protein-like 1
Gene Aliases
Anln, RGD1566097
Chromosome Location
Chr. 17: 66539477 - 66553118 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008771846.1 XP_008770068.1 2 2270

View Full Product Details