Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003551.2 | NP_003542.1 | 2 | 148 | |
XM_005272099.1 | XP_005272156.1 | 1 | 75 | ||
XM_006714716.3 | XP_006714779.1 | 1 | 75 | ||
XM_017009945.1 | XP_016865434.1 | 1 | 75 | ||
GenBank mRNA | AF067724.1 | - | 2 | 118 | |
AK312181.1 | - | 2 | 95 | ||
BC026182.2 | - | 2 | 120 | 5270050 | |
BX490554.1 | - | 2 | 117 | ||
KU178416.1 | - | 1 | 70 | ||
KU178417.1 | - | 1 | 70 | ||
U90450.1 | - | 2 | 94 | ||
Y14992.1 | - | 1 | 84 |