Gene Transcripts
- Gene Symbol
- LOC100911692
- Entrez Gene ID
- 100911692
- Gene Name
- EF-hand calcium-binding domain-containing protein 4A-like
- Gene Aliases
- Chromosome Location
- Chr. 1: 214002860 - 214008970 on Build Rnor_6.0
- Species
- Rattus norvegicus
- Gene Symbol
- Cracr2b
- Entrez Gene ID
- 309112
- Gene Name
- calcium release activated channel regulator 2B
- Gene Aliases
- Efcab4a, RGD1560911
- Chromosome Location
- Chr. 1: 214439766 - 214445876 on Build Rnor_6.0
- UniGene ID
- Rn.13220
- Species
- Rattus norvegicus
View Full Product Details