Gene Transcripts

Gene Symbol
Mms22l
Entrez Gene ID
313108
Gene Name
MMS22-like, DNA repair protein
Gene Aliases
RGD1304693
Chromosome Location
Chr. 5: 38844494 - 38960989 on Build Rnor_6.0
UniGene ID
Rn.23932
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135780.1 NP_001129252.1 14 2501
XM_006237954.2 XP_006238016.1 14 2553

View Full Product Details