Gene Transcripts
- Gene Symbol
- YBX3P1
- Entrez Gene ID
- 440359
- Gene Name
- Y-box binding protein 3 pseudogene 1
- Gene Aliases
- CSDAP1
- Chromosome Location
- Chr. 16: 31567767 - 31569524 on Build GRCh38
- UniGene ID
- Hs.650902
- Species
- Homo sapiens
- Gene Symbol
- YBX3
- Entrez Gene ID
- 8531
- Gene Name
- Y-box binding protein 3
- Gene Aliases
- CSDA, CSDA1, DBPA, ZONAB
- Chromosome Location
- Chr. 12: 10699077 - 10723448 on Build GRCh38
- UniGene ID
- Hs.221889
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001145426.1 |
NP_001138898.1
|
2, 3 |
565 |
|
NM_003651.4 |
NP_003642.3
|
2, 3 |
565 |
|
XM_011520871.2 |
XP_011519173.1
|
2, 3 |
659 |
|
XM_017020122.1 |
XP_016875611.1
|
2, 3 |
659 |
|
XM_017020123.1 |
XP_016875612.1
|
2, 3 |
659 |
|
GenBank mRNA |
AB209896.1 |
- |
2, 3 |
122 |
|
AK314846.1 |
- |
2, 3 |
534 |
|
AU117252.1 |
- |
2, 3 |
565 |
|
BC008801.2 |
- |
2, 3 |
523 |
3634981 |
BC009744.2 |
- |
2, 3 |
518 |
3835480 |
BC015564.1 |
- |
2, 3 |
514 |
4137643 |
BC015913.1 |
- |
2, 3 |
507 |
4563632 |
BC021926.1 |
- |
2, 3 |
516 |
4551107 |
M24069.1 |
- |
2, 3 |
532 |
|
X72712.1 |
- |
2, 3 |
543 |
|
X95325.1 |
- |
2, 3 |
505 |
|
View Full Product Details