Gene Transcripts

Gene Symbol
LOC100911130
Entrez Gene ID
100911130
Gene Name
mitochondrial import inner membrane translocase subunit Tim17-B-like
Gene Aliases
Chromosome Location
Chr. X: 15340557 - 15347674 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003752008.2 XP_003752056.1 232

Gene Symbol
Timm17b
Entrez Gene ID
317374
Gene Name
translocase of inner mitochondrial membrane 17b
Gene Aliases
RGD1561744
Chromosome Location
Chr. X: 15441369 - 15448452 on Build Rnor_6.0
UniGene ID
Rn.17528
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108249.1 NP_001101719.1 3 198

View Full Product Details