Gene Transcripts

Gene Symbol
Utp14a
Entrez Gene ID
317579
Gene Name
UTP14, U3 small nucleolar ribonucleoprotein, homolog A (yeast)
Gene Aliases
Chromosome Location
Chr. X: 135092123 - 135117409 on Build Rnor_6.0
UniGene ID
Rn.16331
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014113.2 NP_001014135.2 12 1653
GenBank mRNA BC088326.1 - 2 813
BC099799.1 - 12 1653 7936638

View Full Product Details