Gene Transcripts

Gene Symbol
Upf2
Entrez Gene ID
361271
Gene Name
UPF2 regulator of nonsense transcripts homolog (yeast)
Gene Aliases
Chromosome Location
Chr. 17: 76176379 - 76287246 on Build Rnor_6.0
UniGene ID
Rn.106291
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108421.1 NP_001101891.1 5 1501

View Full Product Details