Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001009704.1 | NP_001009704.1 | 4 | 2045 | |
XM_008772638.1 | XP_008770860.1 | 4 | 2069 | ||
XM_008772639.1 | XP_008770861.1 | 4 | 2069 | ||
GenBank mRNA | AY043227.1 | - | 4 | 2045 |