Gene Transcripts
- Gene Symbol
- Phactr3
- Entrez Gene ID
- 362284
- Gene Name
- phosphatase and actin regulator 3
- Gene Aliases
- Scapin1, scapinin
- Chromosome Location
- Chr. 3: 173798542 - 173871386 on Build Rnor_6.0
- UniGene ID
- Rn.27183
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_214459.2 |
NP_999624.1
|
11 |
1506 |
|
GenBank mRNA |
AY500157.1 |
- |
11 |
1506 |
|
View Full Product Details