Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001321136.1 | NP_001308065.1 | 13 | 1565 | |
NM_001321137.1 | NP_001308066.1 | 13 | 1162 | ||
NM_003978.4 | NP_003969.2 | 12 | 1346 | ||
XM_006720737.2 | XP_006720800.1 | 12 | 1078 | ||
XM_011522163.1 | XP_011520465.1 | 13 | 1364 | ||
XM_011522164.1 | XP_011520466.1 | 13 | 1032 | ||
XM_011522165.2 | XP_011520467.1 | 12 | 887 | ||
XM_011522166.1 | XP_011520468.1 | 13 | 1398 | ||
XM_011522168.2 | XP_011520470.1 | 13 | 1364 | ||
GenBank mRNA | AB451310.1 | - | 12 | 857 | |
AB451440.1 | - | 12 | 857 | ||
AF038603.1 | - | 12 | 857 | ||
AK308462.1 | - | 13 | 1196 | ||
AK309856.1 | - | 13 | 1528 | ||
BC008602.1 | - | 12 | 1218 | 4180398 | |
KU178458.1 | - | 12 | 857 | ||
U94778.1 | - | 12 | 1073 |