Gene Transcripts

Gene Symbol
Ly49i9
Entrez Gene ID
494205
Gene Name
Ly49 inhibitory receptor 9
Gene Aliases
Chromosome Location
Chr. : - on Build
UniGene ID
Rn.160080
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009496.1 NP_001009496.1 4 444
GenBank mRNA AY649839.1 - 4 444

Gene Symbol
LOC497796
Entrez Gene ID
497796
Gene Name
hypothetical protein LOC497796
Gene Aliases
Chromosome Location
Chr. 4: 164174739 - 164211869 on Build Rnor_6.0
UniGene ID
Rn.160080
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001111310.1 NP_001104780.1 437
XM_006237126.2 XP_006237188.1 4 487
GenBank mRNA BC093399.1 - 437

View Full Product Details