Gene Transcripts

Gene Symbol
Ly49i7
Entrez Gene ID
494210
Gene Name
immunoreceptor Ly49i7
Gene Aliases
Chromosome Location
Chr. 4: 162712736 - 162733947 on Build Rnor_6.0
UniGene ID
Rn.129028
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009500.1 NP_001009500.1 1 51
XM_008763320.1 XP_008761542.1 2 1148
GenBank mRNA AY659934.1 - 1 51

Gene Symbol
LOC100911272
Entrez Gene ID
100911272
Gene Name
killer cell lectin-like receptor 5-like
Gene Aliases
Chromosome Location
Chr. 4: 165347731 - 165366543 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008763440.1 XP_008761662.1 2 907

View Full Product Details