Gene Transcripts

Gene Symbol
Dtx3l
Entrez Gene ID
498089
Gene Name
deltex 3 like, E3 ubiquitin ligase
Gene Aliases
RGD1565144
Chromosome Location
Chr. 11: 67758011 - 67767622 on Build Rnor_6.0
UniGene ID
Rn.1971
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109053.1 NP_001102523.1 3 1675
XM_006248414.2 XP_006248476.1 3 1590

View Full Product Details