Gene Transcripts

Gene Symbol
Ifna5
Entrez Gene ID
690894
Gene Name
interferon alpha family, gene 5
Gene Aliases
Chromosome Location
Chr. 5: 106951822 - 106952908 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001076062.3 XP_001076062.1 1 685

View Full Product Details