Gene Transcripts

Gene Symbol
Pwp2
Entrez Gene ID
690297
Gene Name
PWP2 periodic tryptophan protein homolog (yeast)
Gene Aliases
Chromosome Location
Chr. 20: 11228892 - 11243204 on Build Rnor_6.0
UniGene ID
Rn.208019
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168653.1 NP_001162124.1 10 1166
XM_006256257.2 XP_006256319.1 10 1777

View Full Product Details