Gene Transcripts

Gene Symbol
Srp9
Entrez Gene ID
690345
Gene Name
signal recognition particle 9
Gene Aliases
Chromosome Location
Chr. 13: 100551307 - 100559447 on Build Rnor_6.0
UniGene ID
Rn.7477
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001126095.1 NP_001119567.1 2 277

View Full Product Details