Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005707.1 | NP_005698.1 | 4, 5 | 1397 | |
GenBank mRNA | AF083930.1 | - | 4, 5 | 542 | |
AK096970.1 | - | 4, 5 | 1397 | ||
AK225816.1 | - | 4, 5 | 1364 | ||
BC016992.2 | - | 2, 3 | 327 | ||
BC092464.1 | - | 6, 7 | 721 | ||
BC131704.1 | - | 5, 6 | 699 | ||
BT007395.1 | - | 2, 3 | 217 |