Gene Transcripts

Gene Symbol
ONECUT3
Entrez Gene ID
390874
Gene Name
one cut homeobox 3
Gene Aliases
OC3
Chromosome Location
Chr. 19: 1753663 - 1775445 on Build GRCh38
UniGene ID
Hs.654347
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080488.1 NP_001073957.1 1 1021

View Full Product Details