Gene Transcripts

Gene Symbol
SPRY1
Entrez Gene ID
10252
Gene Name
sprouty RTK signaling antagonist 1
Gene Aliases
hSPRY1
Chromosome Location
Chr. 4: 123396791 - 123403760 on Build GRCh38
UniGene ID
Hs.744854
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001258038.1 NP_001244967.1 2 256
XM_005262686.1 XP_005262743.1 2 260
GenBank mRNA AL602431.1 - 2 257
DA765300.1 - 2 227

View Full Product Details