Gene Transcripts

Gene Symbol
Suclg2
Entrez Gene ID
20917
Gene Name
succinate-Coenzyme A ligase, GDP-forming, beta subunit
Gene Aliases
AF171077, AW556404, D6Wsu120e, SCS-betaG
Chromosome Location
Chr. 6: 95473009 - 95718846 on Build GRCm38
UniGene ID
Mm.371585
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011507.3 NP_035637.2 3 370
GenBank mRNA AF058956.1 - 2 212
AK042987.1 - 3 325
AK045176.1 - 3 323
AK156263.1 - 3 325
AK167136.1 - 3 325
AK167339.1 - 3 609
AK172561.1 - 3 325
AK172633.1 - 3 325
BC043312.1 - 1 65
BC054425.1 - 2 123
BC080781.1 - 3 325 30470296
BI985474.1 - 3 362
EU234009.1 - 3 277

View Full Product Details