Gene Transcripts

Gene Symbol
Map9
Entrez Gene ID
213582
Gene Name
microtubule-associated protein 9
Gene Aliases
5033421J10Rik, 5330427D05Rik, ASAP, Mtap9
Chromosome Location
Chr. 3: 82358028 - 82395268 on Build GRCm38
UniGene ID
Mm.271269
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081230.1 NP_001074699.1 14 2108
GenBank mRNA AK147287.1 - 14 2051
AK147416.1 - 14 2051
BC051460.1 - 6 634

View Full Product Details