Gene Transcripts

Gene Symbol
Vax1
Entrez Gene ID
22326
Gene Name
ventral anterior homeobox 1
Gene Aliases
Chromosome Location
Chr. 19: 59164480 - 59170029 on Build GRCm38
UniGene ID
Mm.23801
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009501.1 NP_033527.1 2 359
GenBank mRNA AF064554.1 - 2 359
BC111818.1 - 2 358 40044698

View Full Product Details