Gene Transcripts

Gene Symbol
Tbc1d8b
Entrez Gene ID
245638
Gene Name
TBC1 domain family, member 8B
Gene Aliases
4921505D17Rik, 9030605E16Rik
Chromosome Location
Chr. X: 139684960 - 139754221 on Build GRCm38
UniGene ID
Mm.275869
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081499.2 NP_001074968.1 7 1266
GenBank mRNA BC147581.1 - 7 1129
BC151057.1 - 7 1129 9087970

View Full Product Details