Gene Transcripts

Gene Symbol
UTP14A
Entrez Gene ID
10813
Gene Name
UTP14A small subunit processome component
Gene Aliases
NYCO16, SDCCAG16, dJ537K23.3
Chromosome Location
Chr. X: 129906121 - 129929762 on Build GRCh38
UniGene ID
Hs.458598
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166221.1 NP_001159693.1 5 368
NM_006649.3 NP_006640.2 5 368
XM_017029237.1 XP_016884726.1 5 302
XM_017029238.1 XP_016884727.1 5 360
GenBank mRNA AK291918.1 - 5 335
AK298578.1 - 5 337
AK304563.1 - 4 233
BC001149.1 - 5 304 3355662
BC009649.1 - 5 306 3459192
BC014987.1 - 5 296 4901477
BP353997.1 - 5 366
BU942896.1 - 5 370

View Full Product Details